missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis C Virus E2 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-62654AF647
This item is not returnable.
View return policy
Description
Hepatitis C Virus E2 Polyclonal antibody specifically detects Hepatitis C Virus E2 in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Hepatitis C Virus E2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 647 | |
| HCV E4, HEPC | |
| Recombinant Hepatitis C Virus E2 (genotype 1a) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction