missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis C Virus E2 Antibody, Janelia Fluor™ 585, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12746JF585
This item is not returnable.
View return policy
Description
Hepatitis C Virus E2 Polyclonal antibody specifically detects Hepatitis C Virus E2 in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Hepatitis C Virus E2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Western Blot | |
| Janelia Fluor 585 | |
| HCV E4, HEPC | |
| Produced in rabbits immunized with purified, recombinant Hepatitis C Virus E2 (Accession#: NP_751921.1; Glu1-Glu278). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction