missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hepatitis C Virus E2 Monoclonal Antibody (I76H)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA17367
This item is not returnable.
View return policy
Description
The MA1-7367 antibody reacts with Hepatitis C virus E2. This antibody is positive against genotypes 1a and 1b. MA1-7367 has been successfully used in ELISA, Immunofluorescence and Western blot applications. The MA1-7367 antibody was raised against HCV E2 protein.
Hepatitis C Virus is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha. Hepatitis C E1 is a virus envelope glycoprotein which forms a heterodimer with the E2 protein.
Specifications
| Hepatitis C Virus E2 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot, Immunocytochemistry | |
| I76H | |
| Unconjugated | |
| HCV E4; HepC | |
| E2 protein. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction