Learn More
Invitrogen™ Hepatitis C Virus NS4 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA173108
Description
The PA1-73108 antibody reacts with Hepatitis C virus NS4 antigen. PA1-73108 has been successfully used in ELISA, and Western blot applications. The PA1-73108 antibody was raised against recombinant NS4 (genotype 1a). Recommended short term, less than 6 months, storage is at 2-8°C. For longer term storage, aliquot and freeze.
HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.
Specifications
| Hepatitis C Virus NS4 | |
| Polyclonal | |
| Unconjugated | |
| HCV; HCV NS4; Hepatitis C Virus; HepC; HepC NS4 | |
| Recombinant NS4 (genotype 1a). | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 5.0 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Goat | |
| IgG fraction | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.