missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis G Virus Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93584AF647
This item is not returnable.
View return policy
Description
Hepatitis G Virus Polyclonal antibody specifically detects Hepatitis G Virus in Human, Virus samples. It is validated for Western Blot
Specifications
| Hepatitis G Virus | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Human, Virus | |
| Purified |
| Western Blot | |
| Alexa Fluor 647 | |
| Hepatitis GB virus C, HGV | |
| Reacts with residues 2268-2276 [CDKCEARQE] of HGV. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction