Learn More
Hepatitis G Virus Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93584APC
This item is not returnable.
View return policy
Description
Hepatitis G Virus Polyclonal antibody specifically detects Hepatitis G Virus in Human, Virus samples. It is validated for Western Blot
Specifications
| Western Blot | |
| Reacts with residues 2268-2276 [CDKCEARQE] of HGV. | |
| RUO | |
| Store at 4C in the dark. |