Learn More
Hepatocyte Specific Antigen Antibody (HSA133), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34644IR
This item is not returnable.
View return policy
Description
Hepatocyte Specific Antigen Monoclonal specifically detects Hepatocyte Specific Antigen in Human samples. It is validated for ELISA.
Specifications
| Hepatocyte Specific Antigen | |
| Monoclonal | |
| DyLight 755 | |
| Hepatocyte, HSA | |
| SK-H1A9-2 human hepatocellular carcinoma cells | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry, Immunofluorescence | |
| HSA133 | |
| Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| monoclonal antibody HSA133 stains human liver canaliculi and a subset of hepatocellular carcinomas. In frozen sections, it stains liver canaliculi strongly and may be used as a marker of this hepatic substructure. Cell preparations of hepatocellular carcinoma biopsies and cell lines are found to bind this monoclonal antibody on the cell surface. HSA133 strongly stains liver canaliculi and hepatic carcinoma cells using frozen sections or paraformaldehyde fixed cell preparations. | |
| Store at 4C in the dark. | |
| IgG2b κ |