missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatocyte Specific Antigen Antibody (HSA133), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34644JF646
This item is not returnable.
View return policy
Description
Hepatocyte Specific Antigen Monoclonal antibody specifically detects Hepatocyte Specific Antigen in Human samples. It is validated for ELISA
Specifications
| Hepatocyte Specific Antigen | |
| Monoclonal | |
| Janelia Fluor 646 | |
| Hepatocyte, HSA | |
| SK-H1A9-2 human hepatocellular carcinoma cells | |
| 0.1 mL | |
| Cellular Markers | |
| Human | |
| Purified |
| ELISA | |
| HSA133 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction