missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepcidin Antimicrobial Peptide Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-36960
This item is not returnable.
View return policy
Description
Hepcidin Antimicrobial Peptide Polyclonal antibody specifically detects Hepcidin Antimicrobial Peptide in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA
Specifications
| Hepcidin Antimicrobial Peptide | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA | |
| hepcidin antimicrobial peptide, HEPCPutative liver tumor regressor, HFE2Bhepcidin, LEAP-1, LEAP1PLTR, Liver-expressed antimicrobial peptide 1 | |
| Peptide with sequence TSGSVFPQQTGQLAE-C corresponding to N-Terminus (near) according to NP_066998.1. | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 μg/mL, Immunohistochemistry 3.75 μg/mL, Immunohistochemistry-Paraffin 3.75 μg/mL, Peptide ELISA Detection limit 1:16000 | |
| NP_066998.1 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| 57817 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction