missing translation for 'onlineSavingsMsg'
Learn More

Melford HEPES, Free Acid

Product Code. p-200087107
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
250g
500g
50g
5kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018333 250 g 250g
30018336 5 kg 5kg
30018335 500 g 500g
30018334 50 g 50g
30018332 1 kg 1kg
30018331 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30018333 Supplier Melford Supplier No. H75030250.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HEPES, Free Acid (4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid), is a zwitterionic buffer.

HEPES, Free Acid (4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid), is a zwitterionic buffer.

In the life science field, HEPES, Free Acid, is primarily utilized as a buffering agent to maintain a stable pH in various experimental procedures. It is particularly effective in the pH range of 6.8 to 8.2, making it suitable for applications requiring slightly acidic to neutral pH conditions.

HEPES, Free Acid, is commonly used in cell culture media, protein purification, and enzyme assays where a specific pH range needs to be maintained. Its buffering capacity and stability within the desired pH range make it valuable in these applications.

One of the significant advantages of HEPES, Free Acid, is its compatibility with biological systems and enzymes. It exhibits minimal interference with enzymatic activity and other biological processes, ensuring reliable and accurate results.

pka (25°C): 7.5

pH Range (Working): 6.8 - 8.2

Heavy Metals: <0.001% Max

Specifications

Chemical Name or Material HEPES, Free Acid
CAS 7365-45-9
Quantity 250 g
Molecular Formula C8H18N2O4S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.