missing translation for 'onlineSavingsMsg'
Learn More

Hepsulfam, MedChemExpress

Product Code. 16442358
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16442358

Brand: Medchem Express HYU00095

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hepsulfam (NCI 329680; ZINC01574758) is a anticancer agent that shows excellent antileukemic activity with an median IC50 of 0.91 μg/mL in a panel of different tumors.

Hepsulfam (NCI 329680

Specifications

Chemical Name or Material Hepsulfam
CAS 96892-57-8
Color White
Purity Grade Notes Research
Molecular Formula C7H18N2O6S2
Quantity 5 mg
Synonym NCI 329680 ZINC01574758
SMILES O=S(OCCCCCCCOS(=O)(N)=O)(N)=O
Molecular Weight (g/mol) 290.36
Formula Weight 290.36
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.