Learn More
Thermo Scientific™ O2 Control (5 to 90%)
For Use With: Heracell™ VIOS 160i and Forma™ Steri-Cycle™ i160 Incubators
Brand:  Thermo Scientific™ 51901138
Additional Details : Weight : 5.00000kg
Product Code. 15333012
  • € 2640.00 / Each
Estimated Shipment: 04-07-2024
to see stock.
Add to basket
Description
Description
Upgrade your incubators to take advantage of O&subscript_2; control.
Specifications
Specifications
5 to 90% O2 control
LED
Accessory, 5 to 90% O2 Control
Heracell™ VIOS 160i and Forma™ Steri-Cycle™ i160 Incubators
5 to 90%