missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HERC4 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-00104JF549
This item is not returnable.
View return policy
Description
HERC4 Polyclonal antibody specifically detects HERC4 in Human, Mouse samples. It is validated for Western Blot
Specifications
| HERC4 | |
| Polyclonal | |
| Western Blot | |
| DKFZP564G092, EC 6.3.2, EC 6.3.2.-, HECT domain and RCC1-like domain-containing protein 4, hect domain and RLD 4, KIAA1593probable E3 ubiquitin-protein ligase HERC4 | |
| The immunogen recognized by this antibody maps to a region between residue 1007 and 1057 of human hect domain and RLD 4 using the numbering given in entry NP_071362.1 (GeneID 26091). | |
| 0.1 mL | |
| Cell Biology | |
| 26091 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction