Learn More
Invitrogen™ HERC5 Monoclonal Antibody (K29053_7B12), Invitrogen™
Mouse Monoclonal Antibody
€ 493.00 - € 868.00
Specifications
| Antigen | HERC5 |
|---|---|
| Clone | K29053_7B12 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30208956
|
Invitrogen™
MA555796 |
50 μL |
€ 493.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30199542
|
Invitrogen™
MA556176 |
200 μL |
€ 868.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: RESEFALRPT FDLTVRRNHL IEDVLNQLSQ FENEDLRKEL WVSFSGEIGY DLGGVKKEFF YCLFAEMIQP EYGMFMYPEG ASCMWFPVKP KFEKKRYFFF GVLCGLSLFN CNVANLPFPL ALFKKLLDQM PSLEDLKELS PDLGKNLQTL LDDEGDNFEE VFYIHFNVHW DRNDTNLIPN GSSITVNQTN KRDYVSKYIN YIFNDSVKAV YEEFRRGFYK MCDEDIIKLF HPEELKDVIV GNTDYDWKTF EKNARYEPGY NSSHPTIVMF WKAFHKLTLE EKKKFLVFLT GTDRLQMKDL NNMKITFCCP ESWNERDPIR ALTCFSVLFL PKYSTMETVE EALQEAINNN
HERC5 Monoclonal antibody specifically detects HERC5 in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| HERC5 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| CEB1; CEBP1; Cyclin-E-binding protein 1; E3 ISG15--protein ligase HERC5; HECT and RLD domain containing E3 ubiquitin protein ligase 5; HECT domain and RCC1-like domain-containing protein 5; hect domain and RLD 5; HERC5; probable E3 ubiquitin-protein ligase HERC5 | |
| Recombinant human HERC5 (immunogen range is 671-1020). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| HERC5 |
| K29053_7B12 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UII4 | |
| 51191 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.