Learn More
Herka™ Conductivitity Meter with 4 LEDs
Product Code. 10045220
Change view
Click to view available options
Flow Rate:
30 to 40 L/hr.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
10045220 30 to 40 L/hr. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10045220 Supplier Herka Lab-In Supplier No. 10320
  • ONLINE EXCLUSIVE PRICE
    € 547.00
    € 456.87 / Pack of 1
    Save € 90.13
    16% Off
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Includes Wall bracket, 2m hose with kraanwartel, 1m outlet hose, and instructions.
Flow Rate 30 to 40 L/hr.