missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus 1 Antibody (LP13) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-66550
This item is not returnable.
View return policy
Description
Herpes Simplex Virus 1 Monoclonal antibody specifically detects Herpes Simplex Virus 1 in Virus samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunomicroscopy.
Specifications
| Herpes Simplex Virus 1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500, Immunomicroscopy 1:10-1:2000 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Binds to an epitope within an Herpes Simplex Virus 1-specific internal phosphorylated protein of molecular weight 45kD. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunomicroscopy | |
| LP13 | |
| Unconjugated | |
| Herpes simplex virus, Herpes simplex virus 1, Herpes simplex virus type 1, HSV 1, Human herpesvirus 1, Human herpesvirus type 1 | |
| Herpes Simplex Virus 1 - strain HFEM | |
| 0.5 mg | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction