missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus 1 Antibody (T111), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-476AF532
This item is not returnable.
View return policy
Description
Herpes Simplex Virus 1 Monoclonal antibody specifically detects Herpes Simplex Virus 1 in Virus samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence.
Specifications
| Herpes Simplex Virus 1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| 0.1 mL | |
| Recognizes gB antigen of both Herpes Simplex Virus 1 &2 | |
| Store at 4°C in the dark. |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| T111 | |
| ELISA, Immunocytochemistry/Immunofluorescence, Immunocytochemistry | |
| Herpes simplex virus, Herpes simplex virus 1, Herpes simplex virus type 1, HSV 1, Human herpesvirus 1, Human herpesvirus type 1 | |
| Exctract of Herpes Simplex Virus 1 infected VERO (green monkey kidney) cells. | |
| Primary | |
| Virus | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction