Learn More
Herpes Simplex Virus 1 Antibody (T111), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-476MFV450
This item is not returnable.
View return policy
Description
Herpes Simplex Virus 1 Monoclonal antibody specifically detects Herpes Simplex Virus 1 in Virus samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Herpes Simplex Virus 1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Immunocytochemistry | |
| T111 | |
| ELISA, Immunocytochemistry/ Immunofluorescence | |
| Herpes simplex virus, Herpes simplex virus 1, Herpes simplex virus type 1, HSV 1, Human herpesvirus 1, Human herpesvirus type 1 | |
| Exctract of Herpes Simplex Virus 1 infected VERO (green monkey kidney) cells. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2b |