missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus 1 ICP8 Antibody (10A3), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2770AF405
This item is not returnable.
View return policy
Description
Herpes Simplex Virus 1 ICP8 Monoclonal antibody specifically detects Herpes Simplex Virus 1 ICP8 in Virus samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Herpes Simplex Virus 1 ICP8 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 10A3 | |
| Western Blot, Immunohistochemistry | |
| DBP, Herpes simplex virus type 1 DBP, Herpes simplex virus type 1 ICP8, Herpes simplex virus type 1 ICP8 major DNA binding protein, Herpes simplex virus type 1 infected cell protein 8, Herpes simplex virus type 1 major DNA binding protein, Herpes simplex virus type 1 UL29, HSV1 DBP, HSV1 ICP8, HSV1 infected cell protein 8, HSV1 major DNA binding protein, HSV1 UL29, ICP 8, ICP 8 protein, ICP8, Infected cell protein 8, Major DNA binding protein, UL 29, UL29 | |
| Herpes Simplex Virus 1 ICP8 purified from U-35-VERO cells. | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction