Learn More
Herpes Simplex Virus UL42 Antibody (13C9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-2761-0.025mg
 View more versions of this product
Product Code. 18526993
  • € 190.00 / 25µg
Description
Herpes Simplex Virus UL42 Monoclonal antibody specifically detects Herpes Simplex Virus UL42 in Virus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Herpes Simplex Virus UL42
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready
HHV1gp061
Baculovirus-expressed Herpes Simplex Virus DNA polymerase (POL) and POL/UL42 complex.
0.025 mg
Virology Bacteria and Parasites
Virus
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
13C9
Unconjugated
PBS
Mouse
Protein A purified
RUO
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.