missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HERV Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93579UV
This item is not returnable.
View return policy
Description
HERV Polyclonal antibody specifically detects HERV in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| HERV | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| DyLight 350 | |
| endogenous retroviral family W, env(C7), member 1, ENV, envelope glycoprotein, Envelope polyprotein gPr73, enverin, Env-W, ENVW, HERV-7q, HERV7Q, HERV-7q Envelope protein, HERV-tryptophan envelope protein, HERV-W, HERV-W Env glycoprotein, HERV-W envelope protein, HERV-W_7q21.2 provirus ancestral Env polyprotein, HERV-W{7q21.1} provirus ancestral Env polyprotein, HERVWENV, HERV-W-ENV, HERVWenvelope protein, human endogenous retrovirus W envC7-1 envelope protein, Syncytin, syncytin A, Syncytin-1 | |
| Synthetic peptide corresponding to residues 42-50 [QRPGNIDAPC] of the HERV protein. | |
| 0.1 mL | |
| Cell Biology | |
| 30816 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction