missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HERV Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93579F
This item is not returnable.
View return policy
Description
HERV Polyclonal antibody specifically detects HERV in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| HERV | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| FITC | |
| endogenous retroviral family W, env(C7), member 1, ENV, envelope glycoprotein, Envelope polyprotein gPr73, enverin, Env-W, ENVW, HERV-7q, HERV7Q, HERV-7q Envelope protein, HERV-tryptophan envelope protein, HERV-W, HERV-W Env glycoprotein, HERV-W envelope protein, HERV-W_7q21.2 provirus ancestral Env polyprotein, HERV-W{7q21.1} provirus ancestral Env polyprotein, HERVWENV, HERV-W-ENV, HERVWenvelope protein, human endogenous retrovirus W envC7-1 envelope protein, Syncytin, syncytin A, Syncytin-1 | |
| Synthetic peptide corresponding to residues 42-50 [QRPGNIDAPC] of the HERV protein. | |
| 0.1 mL | |
| Cell Biology | |
| 30816 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction