missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HES2, Mouse anti-Human, Clone: 4A5, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00054626-M05.100ug
This item is not returnable.
View return policy
Description
Sequence: LPLLGRENSNCSKLEKADVLEMTVRFLQELPASSWPTAAPLPCDSYREGYSACVARLARVLPACRVLEPAVSARSpecifications
| HES2 | |
| Monoclonal | |
| Unconjugated | |
| hairy and enhancer of split 2 (Drosophila) | |
| bHLHb40 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 4A5 | |
| In 1x PBS, pH 7.4 | |
| NM_019089 | |
| HES2 | |
| HES2 (NP_061962, 41 a.a. ∼ 114 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 54626 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction