missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HES6 Antibody (OTI1D6), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72008UV
This item is not returnable.
View return policy
Description
HES6 Monoclonal specifically detects HES6 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| HES6 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HES6 | |
| Full length recombinant protein of human HES6 (NP_061115) produced in E.coli. | |
| 0.1 mL | |
| 55502.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1D6 | |
| Western Blot | |
| BHLHB41, bHLHb41Class B basic helix-loop-helix protein 41, bHLHc23, C-HAIRY1, Hairy and enhancer of split 6, hairy and enhancer of split 6 (Drosophila), HES-6, transcription cofactor HES-6 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction