Learn More
HES6 Mouse anti-Human, Clone: OTI1H2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF800717
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The HES family members form a complex with TLE, the mammalian homologue of Groucho, and this interaction is mediated by the carboxy terminal WRPW motif of the HES proteins. The HES/TLE complex functions by directly binding to DNA, instead of interfering with activator proteins. Most HES family members, including HES1 and HES5, preferentially bind to the N box (CACNAG) as opposed to the E box (CANNTG). HES2 binds to both N and E box sites, while HES6 does not bind DNA. Rather, HES6 inhibits HES1 activity, thereby promoting transcription. HES1 and HES2 are expressed in a variety of adult and embryonic tissues. HES3 is expressed exclusively in cerebellar Purkinje cells, and HES5 is found solely in the nervous system. HES6 is produced in brain as well as in the limb buds of developing embryos.Specifications
| HES6 | |
| Monoclonal | |
| Unconjugated | |
| HES6 | |
| C-HAIRY1, HES-6, bHLHb41, bHLHc23 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55502 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1H2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q96HZ4 | |
| HES6 | |
| Full length human recombit protein of human HES6 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.