missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HES7 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27417MFV500
This item is not returnable.
View return policy
Description
HES7 Polyclonal antibody specifically detects HES7 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| HES7 | |
| Polyclonal | |
| Western Blot | |
| bHLH factor Hes7, BHLHB37, bHLHb37transcription factor HES-7, Class B basic helix-loop-helix protein 37, Hairy and enhancer of split 7, hairy and enhancer of split 7 (Drosophila), hHes7, SCDO4 | |
| RAENRDGPkMLkPLVEkRc (aa 6-23) | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 84667 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction