missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hex Monoclonal Antibody (OTI3C4)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524881
This item is not returnable.
View return policy
Description
Hex Monoclonal Antibody for Western Blot, ICC/IF
HHEX is a member of the homeobox family of transcription factors, many of which are involved in developmental processes. Expression in specific hematopoietic lineages suggests that this protein may play a role in hematopoietic differentiation. Establishes anterior identity at two levels 1) acts early to enhance canonical WNT-signaling by repressing expression of TLE4 and 2) acts later to inhibit NODAL-signaling by directly targeting NODAL.
Specifications
| Hex | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q03014 | |
| HHEX | |
| Recombinant fragment expressed in E.coli corresponding to amino acids 100-270 of Hex. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| OTI3C4 | |
| Unconjugated | |
| HHEX | |
| hematopoietically expressed homeobox; hematopoietically expressed homeobox, related sequence 2; hematopoietically-expressed homeobox protein Hhex; HEX; Hex1; HHEX; Hhex-rs2; HMPH; homeobox; homeobox protein HEX; Homeobox protein PRH; homeobox, hematopoietically expressed; HOX11L-PEN; mHex; Prh; Prhx; proline-rich homeodomain-containing transcription factor | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3087 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction