missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylcyclotrisiloxane, TRC

Product Code. 17912325
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Unit Size:
250g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17912325 5 g 5g
30132719 250 g 250g
30137103 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17912325 Supplier Toronto Research Chemicals Supplier No. TRCH2943405G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-05-9
Formula Weight 222.056
InChI Formula InChI=1S/C6H18O3Si3/c1-10(2)7-11(3,4)9-12(5,6)8-10/h1-6H3
IUPAC Name 2,2,4,4,6,6-hexamethyl-1,3,5,2,4,6-trioxatrisilinane
Molecular Formula C6 H18 O3 Si3
Molecular Weight (g/mol) 222.46
SMILES C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O1
Synonym Cyclotrisiloxane, hexamethyl- (6CI,8CI,9CI), 2,2,4,4,6,6-Hexamethylcyclotrisiloxane, Dimethylsiloxane cyclic trimer, Hexamethylcyclotrisiloxane, LS 8120, SDK 10
Chemical Name or Material Hexamethylcyclotrisiloxane

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.