Learn More
Hexamethyldisiloxane, 98+%
Product Code. 11448890
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
25mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448890 25 mL 25mL
11423814 100 mL 100mL
11433814 500 mL 500mL
11443814 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11448890 Supplier Thermo Scientific Alfa Aesar Supplier No. A11848.AC
 Restricted Item
In Stock
Add to basket
CAS: 107-46-0 | C6H18OSi2 | 162.379 g/mol
Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule. It is also used to make low-k dielectric materials for the semi-conductor industries by plasma-enhanced chemical vapor deposition(PECVD) method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule. It is also used to make low-k dielectric materials for the semi-conductor industries by plasma-enhanced chemical vapor deposition(PECVD) method.
Solubility
Slightly miscible with water.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, bases and oxygen.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 107-46-0
Molecular Formula C6H18OSi2
MDL Number MFCD00008265
InChI Key UQEAIHBTYFGYIE-UHFFFAOYSA-N
Synonym hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide
PubChem CID 24764
ChEBI CHEBI:78002
IUPAC Name trimethyl(trimethylsilyloxy)silane
SMILES C[Si](C)(C)O[Si](C)(C)C
Specifikationer
Melting Point -67°C
Density 0.764
Boiling Point 100°C to 101°C
Flash Point -2°C
Odor Characteristic
Linear Formula (CH3)3SiOSi(CH3)3
Refractive Index 1.377
Quantity 25 mL
UN Number UN1993
Beilstein 1736258
Solubility Information Slightly miscible with water.
Molecular Weight (g/mol) 162.38
Formula Weight 162.38
Percent Purity ≥98%
Chemical Name or Material Hexamethyldisiloxane
Visa mer Visa mindre
RUO – Research Use Only
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.