missing translation for 'onlineSavingsMsg'
Learn More

Hexanoyl-d11 Glycine, TRC

Product Code. 30179468
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179468 25 mg 25mg
30179600 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30179468 Supplier Toronto Research Chemicals Supplier No. TRCH28120325MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C8 D11 H4 N O3
Molecular Weight (g/mol) 184.28
Synonym N-(1-Oxohexyl)glycine-d11, Caproylglycine-d11, N-Caproylglycine-d11, NSC 224460-d11, n-Hexanoylglycine-d11, N-Hexanoyl-d11-glycine, N-(1-Oxohexyl-2,2,3,3,4,4,5,5,6,6,6-d11)glycine, N-Hexanoyl-d11-glycine, N-Hexanoylglycine-D11 (hexanoyl-D11)
IUPAC Name 2-(2,2,3,3,4,4,5,5,6,6,6-undecadeuteriohexanoylamino)acetic acid
SMILES [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)NCC(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C8H15NO3/c1-2-3-4-5-7(10)9-6-8(11)12/h2-6H2,1H3,(H,9,10)(H,11,12)/i1D3,2D2,3D2,4D2,5D2
Formula Weight 184.1742
Chemical Name or Material Hexanoyl-d11 Glycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.