Learn More
Invitrogen™ Hexanoyl-Lysine adduct Monoclonal Antibody (5D9), APC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545777
Description
A 1:1,000 dilution of MA5-45777 was sufficient for detection of Hexanoyl Lysine adduct in 0.5 μg of Hexanoyl Lysine conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody. Specific for Hexanoyl-Lysine adduct (HEL) modified peptides and proteins. Does not detect free Hexanoyl-Lysine. Does not cross-react with Acrolein, Crotonaldehyde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins.
Hexanoyl-lysine (HEL) is a novel lipid peroxidation biomarker which is derived from the oxidation of omega-6 unsaturated fatty acid. HEL may be used as a biomarker in omega-6 PUFA (polyunsaturated fatty acid) oxidation in living organisms.
Specifications
| Hexanoyl-Lysine adduct | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 5D9 | |
| APC | |
| HEL; HEL Adduct; Hexanoyl-Lys; Hexanoyl-Lys (HEL); Hexanoyl-Lys adduct | |
| Synthetic Hexanoyl modified Keyhole Limpet Hemocyanin (KLH). | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid | |
| Chemical |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.