missing translation for 'onlineSavingsMsg'
Learn More

HEXB Antibody (HEXB/7762) [Janelia Fluor™ 635], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP3-23982JF635

Product Code. 30165763

  • € 672.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HEXB Monoclonal antibody specifically detects HEXB in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HEXB
Monoclonal
Janelia Fluor 635
beta-hexosaminidase subunit beta, Beta-N-acetylhexosaminidase subunit beta, Cervical cancer proto-oncogene 7 protein, EC 3.2.1.52, ENC-1AS, HCC-7, hexosaminidase B (beta polypeptide), Hexosaminidase subunit B, N-acetyl-beta-glucosaminidase subunit beta
Recombinant fragment human HEXB protein (exact sequence is proprietary)
0.1 mL
Neurodegeneration
3074
Store at 4°C in the dark.
IgG2 κ
Immunohistochemistry (Paraffin)
HEXB/7762
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.