missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEXB Antibody (HEXB/7762), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23584-100ug
This item is not returnable.
View return policy
Description
HEXB Monoclonal antibody specifically detects HEXB in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HEXB | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| beta-hexosaminidase subunit beta, Beta-N-acetylhexosaminidase subunit beta, Cervical cancer proto-oncogene 7 protein, EC 3.2.1.52, ENC-1AS, HCC-7, hexosaminidase B (beta polypeptide), Hexosaminidase subunit B, N-acetyl-beta-glucosaminidase subunit beta | |
| Recombinant fragment human HEXB protein (exact sequence is proprietary) | |
| 100 μg | |
| Neurodegeneration | |
| 3074 | |
| Store at 4°C. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| HEXB/7762 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction