missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HEXB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536146
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 63 kDa. Purity is >95% by SDS-PAGE.
Hexosaminidase B (HEXB), also designated beta-hexosaminidase B, is a Hexosaminidase B (HEXB), also designated b-hexosaminidase B, is a tetramer of two b-A and two b-B chains and is found in the lysosomes of cells. Sandhoff disease (SD), also known as GM2-gangliosidosis type II, is caused by mutations in the HEXB gene that affect the b subunit. These mutations disrupt the activity of HEXB and HEXA, which prevents the breakdown of GM2 ganglioside, a fatty material found in the brain, therby rendering both the HEXA and HEXB enzymes deficient. SD is a rare autosomal recessive disorder characterized by an accumulation of GM2 ganglioside, which causes progressive destruction of the central nervous system. Sandhoff disease is similar to Tay-Sachs disease, which is caused by mutations in the HEXA gene, although SD is more severe.
Specifications
| HEXB | |
| Polyclonal | |
| Unconjugated | |
| Hexb | |
| beta-hexosaminidase subunit beta; Beta-hexosaminidase subunit beta chain A; Beta-hexosaminidase subunit beta chain B; Beta-N-acetylhexosaminidase subunit beta; cervical cancer proto-oncogene 7 protein; ENC-1AS; epididymis luminal protein 248; epididymis secretory protein Li 111; HCC7; HCC-7; HEL-248; HEL-S-111; HEXB; hexosaminidase B; hexosaminidase B (beta polypeptide); hexosaminidase subunit B; hexosaminidase subunit beta; N-acetyl-beta-glucosaminidase subunit beta | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3074 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P07686 | |
| Hexb | |
| Synthetic peptide corresponding to amino acids 481-530 of human Hexb. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction