missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEXO Antibody (OTI1B3), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72312CL1
This item is not returnable.
View return policy
Description
HEXO Monoclonal antibody specifically detects HEXO in Human,Monkey,Primate,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot
Specifications
| HEXO | |
| Monoclonal | |
| CoraFluor 1 | |
| 3' exoribonuclease, 3'-5' exonuclease ERI1, 3'EXO, 3'HEXO, EC 3.1, enhanced RNAi three prime mRNA exonuclease homolog 1, Eri-1 homolog, exoribonuclease 1, HEXO, histone mRNA 3' end-specific exonuclease, Histone mRNA 3'-end-specific exoribonuclease, Histone mRNA 3'-exonuclease 1, MGC35395, Protein 3'hExo, THEX1, three prime histone mRNA exonuclease 1,3'-5' exoribonuclease 1, three prime mRNA exonuclease 1 | |
| Full length human recombinant protein of human ERI1(NP_699163) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Monkey, Primate, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| OTI1B3 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 90459 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction