missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexokinase 1 Antibody (7A7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51639
This item is not returnable.
View return policy
Description
Hexokinase 1 Monoclonal specifically detects Hexokinase 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Hexokinase 1 | |
| Monoclonal | |
| Unconjugated | |
| P19367 | |
| HK1 | |
| Purified recombinant fragment of human HK1 expressed in E. Coli (between residues 290-430). [UniProt# P19367] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Ascites |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 7A7 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Brain form hexokinase, EC 2.7.1, EC 2.7.1.1, glycolytic enzyme, hexokinase 1, Hexokinase type I, hexokinase-1, HK I, HK1-ta, HK1-tb, HK1-tc, HKI, HXK1 | |
| Mouse | |
| 102 kDa | |
| 0.1 mL | |
| Cancer | |
| 3098 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction