missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexokinase 1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32166AF532
This item is not returnable.
View return policy
Description
Hexokinase 1 Polyclonal antibody specifically detects Hexokinase 1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Hexokinase 1 | |
| Polyclonal | |
| Western Blot | |
| Brain form hexokinase, EC 2.7.1, EC 2.7.1.1, glycolytic enzyme, hexokinase 1, Hexokinase type I, hexokinase-1, HK I, HK1-ta, HK1-tb, HK1-tc, HKI, HXK1 | |
| The immunogen this antibody was made to, maps to a region between residue 325 to 375 of human Hexokinase 1 using the numbering given in entry NP_000179.2 (GeneID 3098). | |
| 0.1 mL | |
| Cancer | |
| 3098 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction