missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexosaminidase A/HEXA Antibody (20F1), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42629AF594
This item is not returnable.
View return policy
Description
Hexosaminidase A/HEXA Monoclonal specifically detects Hexosaminidase A/HEXA in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| Hexosaminidase A/HEXA | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| HEXA | |
| Recombinant human HEXA (89-529aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Human | |
| IgG2a λ |
| Western Blot, Flow Cytometry, ELISA | |
| 20F1 | |
| Western Blot, Flow Cytometry, ELISA | |
| beta-hexosaminidase subunit alpha, Beta-N-acetylhexosaminidase subunit alpha, EC 3.2.1, EC 3.2.1.52, hexosaminidase A (alpha polypeptide), Hexosaminidase subunit A, MGC99608, N-acetyl-beta-glucosaminidase subunit alpha, TSD | |
| Mouse | |
| Protein A purified | |
| Neuroscience, Signal Transduction | |
| 3073.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction