missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexosaminidase A/HEXA Antibody (20F1), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42629JF646
This item is not returnable.
View return policy
Description
Hexosaminidase A/HEXA Monoclonal antibody specifically detects Hexosaminidase A/HEXA in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| Hexosaminidase A/HEXA | |
| Monoclonal | |
| Janelia Fluor 646 | |
| beta-hexosaminidase subunit alpha, Beta-N-acetylhexosaminidase subunit alpha, EC 3.2.1, EC 3.2.1.52, hexosaminidase A (alpha polypeptide), Hexosaminidase subunit A, MGC99608, N-acetyl-beta-glucosaminidase subunit alpha, TSD | |
| Recombinant human Hexosaminidase A/HEXA (89-529aa) purified from E. coli | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 3073 | |
| Store at 4°C in the dark. | |
| IgG2a λ |
| Western Blot, Flow Cytometry, ELISA | |
| 20F1 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction