missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HEY1 Recombinant Rabbit Monoclonal Antibody (HL2367)
GREENER_CHOICE

Product Code. 18035591
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18035591 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18035591 Supplier Invitrogen™ Supplier No. MA550506

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

HEY1 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

Hairy/enhancer-of-split related with YRPW motif protein 1 belongs to the HEY family and is a Notch Signaling Pathway Protein. It contains one basic helix-loop-helix (bHLH) domain and one Orange domain. It is a downstream effector of Notch signaling which may be required for cardiovascular development. It acts as transcriptional repressor which binds preferentially to the canonical E box sequence 5'-CACGTG-3'. It represses transcription by the cardiac transcriptional activators GATA4 and GATA6. Hey1 is also reported to be expressed during development of the nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEY1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL2367
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene HEY1
Gene Accession No. Q9WV93, Q9Y5J3
Gene Alias AI316788; AI414254; basic helix-loop-helix protein OAF1; basic helix-loop-helix transcription factor; BC8; bHLH protein Hesr-1/Hey1; bHLHb31; cardiovascular basic helix-loop-helix factor 2; cardiovascular helix-loop-helix factor 2; CHF2; CHF-2; class B basic helix-loop-helix protein 31; Hairy and enhancer of split-related protein 1; Hairy/E(spl)-related with YRPW motif 1; hairy/enhancer-of-split related with YRPW motif 1; hairy/enhancer-of-split related with YRPW motif protein 1; hairy-related transcription factor 1; Herp1; Herp2; hes related family bHLH transcription factor with YRPW motif 1; Hesr1; HESR-1; hes-related family bHLH transcription factor with YRPW motif 1; HES-related repressor protein 1; HES-related repressor protein 2; HEY1; hey1 protein; hHRT1; HRT1; HRT-1; id:ibd1292; MGC1274; mHRT1; OAF1; zgc:110572
Gene Symbols HEY1
Host Species Rabbit
Immunogen Recombinant fragment of human HEY1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15213, 155437, 23462
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.