Learn More
HEY2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10597-1-AP-150UL
 View more versions of this product
Product Code. 16807083
  • € 410.00 / 150µL
Description
This antibody is a rabbit polyclonal antibody raised against full length of human HEY2.
Downstream effector of Notch signaling which may be required for cardiovascular development. Transcriptional repressor which binds preferentially to the canonical E box sequence 5′-CACGTG-3′. Represses transcription by the cardiac transcriptional activators GATA4 and GATA6.
Specifications
Specifications
HEY2
Polyclonal
Unconjugated
HEY2
bHLHb32, CHF1, GRIDLOCK, GRL, hCHF1, HERP, HERP1, HESR 2, HESR2, HEY2, hHRT2, HRT 2, HRT2, Protein gridlock homolog
Rabbit
Antigen Affinity Chromatography
RUO
15214, 155430, 23493
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9QUS4, Q9UBP5
HEY2
HEY2 Fusion Protein Ag0913
150 μL
Primary
Human, Mouse, Rat