missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HFE2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119184
This item is not returnable.
View return policy
Description
HFE2 Polyclonal Antibody for Western Blot, IHC (P)
HFE2 is involved in iron metabolism. It may be a component of the signaling pathway which activates hepcidin or it may act as a modulator of hepcidin expression. It could also represent the cellular receptor for hepcidin. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene. Defects in this gene are the cause of hemochromatosis type 2A, also called juvenile hemochromatosis (JH). JH is an early-onset autosomal recessive disorder due to severe iron overload resulting in hypogonadotrophic hypogonadism, hepatic fibrosis or cirrhosis and cardiomyopathy, occurring typically before age of 30.
Specifications
| HFE2 | |
| Polyclonal | |
| Unconjugated | |
| HFE2 | |
| 2310035L15Rik; 5230400G09Rik; AI414844; AI789733; DL M; DL-M; Fidgetin; haemojuvelin; HEMOCHROMATOSIS; hemochromatosis type 2 (juvenile); hemochromatosis type 2 (juvenile) (human homolog); hemochromatosis type 2 protein; Hemochromatosis type 2 protein homolog; hemojuvelin; Hemojuvelin BMP coreceptor; HFE 2; HFE2; HFE2A; HJV; JH; MGC23953; OTTHUMP00000059680; repulsive guidance molecule c; RGM C; RGM domain family member C; RGMC | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 148738, 310681 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q6ZVN8, Q8N7M5 | |
| HFE2 | |
| Recombinant protein Hemojuvelin. The antigen corresponds to amino acid range 29-401 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction