Learn More
Invitrogen™ HFM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109810
Description
Immunogen sequence: MLKSNDCLFS LENLFFEKPD EVENHPDNEK SLDWFLPPAP LISEIPDTQE LEEELESHKL LGQEKRPKML TSNLKITNED TNYISLTQKF QFAFPSDKYE QDDLNLEGVG NNDLSHVAGK LTYASQKYKN HIGTEIAPEK SVPDDTKLVN FAEDKGESTS VFRKRLFKIS DNIHGSAYSN DNELDSHIGS VKIVQTEMNK GKSRNYSNSK QKFQYSANVF TANNAFSASE IGEGMFKAPS.
The protein encoded by this gene is thought to be an ATP-dependent DNA helicase and is expressed mainly in germ-line cells. Defects in this gene are a cause of premature ovarian failure 9 (POF9).
Specifications
| HFM1 | |
| Polyclonal | |
| Unconjugated | |
| HFM1 | |
| A330009G12Rik; Gm1046; helicase; helicase-like protein HFM1; Hfm1; HFM1, ATP dependent DNA helicase homolog; HFM1, ATP-dependent DNA helicase homolog; HFM1, ATP-dependent DNA helicase homolog (S. cerevisiae); Mer3; POF9; probable ATP-dependent DNA helicase HFM1; RP11-539G11.1; SEC3D1; SEC63 domain containing 1; SEC63 domain-containing protein 1; SEC63D1; Si-11; Si-11-6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 164045, 330149, 690161 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.55 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A2PYH4, D3Z4R1 | |
| HFM1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human HFM1 (NP_001017975.4). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.