missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HGF Antibody (016) [FITC], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26881F
This item is not returnable.
View return policy
Description
HGF Monoclonal antibody specifically detects HGF in Human samples. It is validated for ELISA
Specifications
| HGF | |
| Monoclonal | |
| FITC | |
| deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, HGFB, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SFhepatopoeitin-A | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human HGF (RefSeq#: NP_000592.3; Met1-Ser728). | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 3082 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 016 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction