missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HGF Antibody (OTI1D2), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70884R
This item is not returnable.
View return policy
Description
HGF Monoclonal specifically detects HGF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| HGF | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HGF | |
| Recombinant protein fragment corresponding to amino acids 200-494 of human HGF (NP_000592) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI1D2 | |
| Western Blot, Immunohistochemistry | |
| deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, HGFB, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SFhepatopoeitin-A | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cell Cycle and Replication | |
| 3082.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction