missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HGF Antibody (OTI1D2), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70884PCP
This item is not returnable.
View return policy
Description
HGF Monoclonal antibody specifically detects HGF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| HGF | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI1D2 | |
| Western Blot, Immunohistochemistry | |
| deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A, scatter factor), Hepatopoeitin-A, HGFB, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SFhepatopoeitin-A | |
| Human recombinant protein fragment corresponding to amino acids 200-494 of human HGF(NP_000592) produced in E.coli. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 3082 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction