missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HGF Chimeric Recombinant Human Monoclonal Antibody (mh010)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535993
This item is not returnable.
View return policy
Description
HGF Chimeric Recombinant Monoclonal Antibody for Neu
Hepatocyte growth factor regulates cell growth, cell motility, and morphogenesis by activating a tyrosine kinase signaling cascade after binding to the proto-oncogenic c-Met receptor. Hepatocyte growth factor is secreted by mesenchymal cells and acts as a multi-functional cytokine on cells of mainly epithelial origin. Its ability to stimulate mitogenesis, cell motility, and matrix invasion gives it a central role in angiogenesis, tumorogenesis, and tissue regeneration. It is secreted as a single inactive polypeptide and is cleaved by serine proteases into a 69-kDa alpha-chain and 34-kDa beta-chain. A disulfide bond between the alpha and beta chains produces the active, heterodimeric molecule. The protein belongs to the plasminogen subfamily of S1 peptidases but has no detectable protease activity. Alternative splicing of this gene produces multiple transcript variants encoding different isoforms.
Specifications
| HGF Chimeric | |
| Recombinant Monoclonal | |
| 1.4 mg/mL | |
| PBS with no preservative | |
| P14210 | |
| Hgf | |
| Recombinant Human HGF/Hepatocyte Growth Factor protein. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Neutralization | |
| mh010 | |
| Unconjugated | |
| Hgf | |
| C230052L06Rik; DFNB39; Fibroblast derived tumor cytotoxic factor; fibroblast-derived tumor cytotoxic factor; F-TCF; hepatocyte growth factor; Hepatocyte growth factor (hepapoietin A; hepatocyte growth factor (hepapoietin A; scatter factor); Hepatocyte growth factor alpha chain; Hepatocyte growth factor beta chain; Hepatopoeitin A; hepatopoeitin-A; HepatopoietinA; hepatopoietin-A; Hgf; HGF/SF; HGFB; HPTA; Lung fibroblast derived mitogen; lung fibroblast-derived mitogen; NK1; NK2; Scatter factor; scatter factor); SF; SF/HGF | |
| Human | |
| Protein A | |
| RUO | |
| 3082 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction