missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HGFR/c-MET Antibody (17) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 190.00 - € 351.00
Specifications
| Antigen | HGFR/c-MET |
|---|---|
| Clone | 17 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18622278
|
Novus Biologicals
NBP2-50171-0.025ml |
0.025 mL |
€ 190.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609007
|
Novus Biologicals
NBP2-50171 |
0.1 mL |
€ 351.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HGFR/c-MET Monoclonal antibody specifically detects HGFR/c-MET in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOFSpecifications
| HGFR/c-MET | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, CyTOF | |
| Unconjugated | |
| Mouse | |
| Cancer, Cancer Stem Cells, Cell Cycle and Replication, Cellular Markers, Oncogenes, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| PBS | |
| 4233 | |
| IgG1 κ | |
| Protein A purified |
| 17 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AUTS9, c-Met, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF receptor, HGF/SF receptor, HGFR, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met | |
| Bacterially expressed human HGFR/c-MET alpha chain | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title