missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HGFR/c-MET Antibody (8), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50172APC
This item is not returnable.
View return policy
Description
HGFR/c-MET Monoclonal specifically detects HGFR/c-MET in Human samples. It is validated for Flow Cytometry.
Specifications
| HGFR/c-MET | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| MET | |
| Bacterially expressed human HGFR/c-MET alpha chain | |
| 0.1 mL | |
| Cancer, Cancer Stem Cells, Cell Cycle and Replication, Cellular Markers, Oncogenes, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 4233 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 8 | |
| Flow Cytometry | |
| AUTS9, c-Met, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF receptor, HGF/SF receptor, HGFR, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction