missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HGK/MAP4K4 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74630B
This item is not returnable.
View return policy
Description
HGK/MAP4K4 Polyclonal antibody specifically detects HGK/MAP4K4 in Human samples. It is validated for Western Blot
Specifications
| HGK/MAP4K4 | |
| Polyclonal | |
| Western Blot | |
| EC 2.7.11, EC 2.7.11.1, FLH21957, FLJ10410, hepatocyte progenitor kinase-like/germinal center kinase-like kinase, HGKFLJ20373, HPK/GCK-like kinase HGK, KIAA0687, MAPK/ERK kinase kinase kinase 4, MEK kinase kinase 4, MEKKK 4, mitogen-activated protein kinase kinase kinase kinase 4, Nck-interacting kinase, NIKFLJ90111 | |
| The immunogen recognized by this antibody maps to a region between residue 850 and 900 of human MAPK/ERK kinase kinase kinase 4 (hepatocyte progenitor kinase-like/germinal center kinase-like kinase) using the numbering given in entry AAO32626.1 (GeneID 94 | |
| 0.1 mL | |
| Phospho Specific, Protein Kinase | |
| 9448 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction